Diisobutylamine hydrochloride 95%

Supplier: Ambeed
Warning

A536413-1G A536413-5G
76805-332EA 9 USD
76805-332 76805-334
Diisobutylamine hydrochloride 95%
Diisobutylamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₉N
MDL Number: MFCD00060151
CAS Number: 18251-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR