4-Methoxyindole-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A534637-100MG
A534637-10G
A534637-1G
A534637-250MG
A534637-25G
A534637-5G
76805-306EA
5.31
USD
76805-306
76805-308
76805-310
76805-312
76805-314
76805-316
4-Methoxyindole-2-carboxylic acid 98%
4-Methoxyindole-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₃ MW: 191.19 g/mol |
MDL Number:
MFCD02664458 CAS Number: 103260-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...