9-Benzylcarbazole-3,6-dicarboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 9-Benzyl-9H-carbazole-3,6-dicarbaldehyde, 9-Benzyl-3,6-diformylcarbazole

A531858-100MG A531858-1G A531858-250MG
76805-284EA 59 USD
76805-284 76805-286 76805-288
9-Benzylcarbazole-3,6-dicarboxaldehyde 98%
9-Benzylcarbazole-3,6-dicarboxaldehyde

Precaución: for Research Use Only

Formula: C₂₁H₁₅NO₂
MW: 313.36 g/mol
MDL Number: MFCD08276316
CAS Number: 200698-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR