Ethyl 2-(2-methyl-1H-indol-3-yl)acetate 97%

Supplier: Ambeed
Warning

A530948-100MG A530948-1G A530948-250MG
76805-272EA 56.23 USD
76805-272 76805-274 76805-276
Ethyl 2-(2-methyl-1H-indol-3-yl)acetate 97%
Ethyl 2-(2-methyl-1H-indol-3-yl)acetate

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₂
MW: 217.26 g/mol
MDL Number: MFCD02093647
CAS Number: 21909-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR