Disodium-1,6-naphthalenedisulfonate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,6-Naphthalenedisulphonic acid Disodium salt, Sodium naphthalene-1,6-disulphonate
A529998-100G
A529998-25G
A529998-500G
76805-266EA
14.86
USD
76805-266
76805-268
76805-270
Disodium-1,6-naphthalenedisulfonate 97%
Disodium-1,6-naphthalenedisulfonate
Precaución: for Research Use Only
Formula:
C₁₀H₈O₆S₂ MW: 332.25 g/mol |
MDL Number:
MFCD00021522 CAS Number: 1655-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...