3-(4-Thiomorpholinylcarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-(Thiomorpholine-4-carbonyl)phenyl)boronic acid

A529607-1G A529607-250MG A529607-5G
76805-260EA 166.57 USD
76805-260 76805-262 76805-264
3-(4-Thiomorpholinylcarbonyl)phenylboronic acid 98%
3-(4-Thiomorpholinylcarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄BNO₃S
MW: 251.1100 g/mol
MDL Number: MFCD06659887
CAS Number: 850567-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR