N-(p-Toluenesulfonyl)-3-pyrroline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Tosyl-2,5-dihydro-1H-pyrrole, N-Tosyl-3-pyrroline
A528804-1G
A528804-25G
A528804-5G
76805-256EA
33
USD
76805-256
76805-258
77880-145
N-(p-Toluenesulfonyl)-3-pyrroline 98%
N-(p-Toluenesulfonyl)-3-pyrroline
Precaución: for Research Use Only
Formula:
C₁₁H₁₃NO₂S |
MDL Number:
MFCD01321189 CAS Number: 16851-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...