Guanidine sulfamate 90%

Supplier: Ambeed
Warning

A528067-25G A528067-5G
76805-254EA 17 USD
76805-254 77880-144
Guanidine sulfamate 90%
Guanidine sulfamate

Precaución: for Research Use Only

Formula: CH₅N₃
MDL Number: MFCD00060261
CAS Number: 50979-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR