(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-(+)-N-(4-toluenesulphonyl)-1,2-diphenylethylenediamine, (S,S)-N-(p-Toluenesulphonyl)-1,2-diphenylethylenediamine, (S,S)-TsDPEN, N-((1S,2S)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulphonamide

A526827-10G A526827-1G A526827-250MG A526827-5G A526827-25G
76805-238EA 79.57 USD
76805-238 76805-240 76805-242 76805-244 77602-244
(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide 98%
(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide

Precaución: for Research Use Only

Formula: C₂₁H₂₂N₂O₂S
MW: 366.48 g/mol
MDL Number: MFCD03095684
CAS Number: 167316-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR