N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Methyl ((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)-L-valinate hydrochloride, N-(2'-Cyanobiphenyl-4-ylmethyl) L-valine methyl ester monohydrochloride

A526618-100G A526618-25G A526618-500G A526618-5G
76805-222EA 41 USD
76805-222 76805-224 76805-226 76805-228
N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride 97%
N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₂O₂
MDL Number: MFCD08460094
CAS Number: 482577-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR