Dorzolamide hydrochloride 98%

Supplier: Ambeed
Warning

A525496-100MG A525496-10MG A525496-1G A525496-1MG A525496-250MG A525496-50MG A525496-5G
76805-200EA 49 USD
76805-200 76805-202 76805-204 76805-206 76805-208 76805-210 76805-212
Dorzolamide hydrochloride 98%
Dorzolamide hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₄S₃
MDL Number: MFCD00884659
CAS Number: 130693-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR