Methyl 2-bromobut-2-enoate 95%

Supplier: Ambeed
Danger

A525032-1G A525032-250MG A525032-5G
76805-196EA 130.5 USD
76805-196 76805-198 77572-672
Methyl 2-bromobut-2-enoate 95%
Methyl 2-bromobut-2-enoate

Precaución: for Research Use Only

Formula: C₅H₇BrO₂
MDL Number: MFCD00042891
CAS Number: 17642-18-1
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR