Diethylene glycol diacrylate 75% stabilized

Supplier: Ambeed
Danger

Synonyms: Diethyleneglycoldiacrylate

Stabilization: Stabilised with hydroquinone (HQ)

A523645-25G A523645-5G
76805-192EA 27.59 USD
76805-192 76805-194
Diethylene glycol diacrylate 75% stabilized
Diethylene glycol diacrylate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₅
MW: 214.22 g/mol
MDL Number: MFCD00014939
CAS Number: 4074-88-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR