Ethyl-2-fluorobenzoate 98%

Supplier: Ambeed
Warning

A522244-10G A522244-25G A522244-5G A522244-100G
76805-172EA 6 USD
76805-172 76805-174 76805-176 77669-312
Ethyl-2-fluorobenzoate 98%
Ethyl-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD00039214
CAS Number: 443-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR