(R)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide 95%

Supplier: Ambeed
Danger

A520900-1G A520900-250MG
76805-156EA 165 USD
76805-156 76805-158
(R)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide 95%
(R)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide

Precaución: for Research Use Only

Formula: C₉H₉FN₄O₅
MDL Number: MFCD00155642
CAS Number: 132055-99-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR