Furazolidone 99%

Supplier: Ambeed
Warning

Synonyms: 3-(((5-Nitrofuran-2-yl)methylene)amino)oxazolidin-2-one, 3-[(5-Nitrofurfurylidene)amino]-2-oxazolidinone

A515238-1000G A515238-100G A515238-250MG A515238-25G A515238-500G
76805-110EA 432.85 USD
76805-110 76805-112 76805-114 76805-116 76805-118
Furazolidone 99%
Furazolidone

Precaución: for Research Use Only

Formula: C₈H₇N₃O₅
MW: 225.16 g/mol
MDL Number: MFCD00910550
CAS Number: 67-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR