1-[5-(Trifluoromethyl)pyridin-2-yl]homopiperazine 97+%

Supplier: Ambeed
Warning

A514276-1G A514276-250MG A514276-5G
76805-102EA 35 USD
76805-102 76805-104 76805-106
1-[5-(Trifluoromethyl)pyridin-2-yl]homopiperazine 97+%
1-[5-(Trifluoromethyl)pyridin-2-yl]homopiperazine

Precaución: for Research Use Only

Formula: C₁₁H₁₄F₃N₃
MDL Number: MFCD00661783
CAS Number: 306934-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR