5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide 98%

Supplier: Ambeed
Warning

Synonyms: Iohexol, N1,N3-Bis(2,3-dihydroxypropyl)-5-(N-(2,3-dihydroxypropyl)acetamido)-2,4,6-triiodoisophthalamide, 5-[Acetyl(2,3-dihydroxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide, N,N'-Bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodoisophthalamide

A514163-100MG A514163-250MG A514163-25MG A514163-1G A514163-5G
76805-096EA 8 USD
76805-096 76805-098 76805-100 77669-284 77669-286
5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide 98%
5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide

Precaución: for Research Use Only

Formula: C₁₉H₂₆I₃N₃O₉
MDL Number: MFCD00077732
CAS Number: 66108-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR