1-(4-Chlorophenyl)cyclopropanecarbonitrile 98%

Supplier: Ambeed
Warning

A514024-1G A514024-25G A514024-5G
76805-086EA 7 USD
76805-086 76805-088 76805-090
1-(4-Chlorophenyl)cyclopropanecarbonitrile 98%
1-(4-Chlorophenyl)cyclopropanecarbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₈ClN
MDL Number: MFCD00019211
CAS Number: 64399-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR