(±)-γ-Heptalactone 98%

Supplier: Ambeed
Warning

Synonyms: 5-Propyldihydrofuran-2(3H)-one, gamma-Enanthonolactone, gamma-Heptalactone, gamma-Heptanolactone, (±)-Heptan-4-olide

A513915-1000G A513915-100G A513915-25G A513915-500G A513915-5G
76805-076EA 127 USD
76805-076 76805-078 76805-080 76805-082 76805-084
(±)-γ-Heptalactone 98%
(±)-γ-Heptalactone

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
MDL Number: MFCD00036498
CAS Number: 105-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR