Dimethyl succinate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
DBE-4 dibasic ester, Succinic acid dimethyl ester, 1,4-Butanedioic acid dimethyl ester
A512762-25G
A512762-1000G
A512762-500G
A512762-100G
76805-070EA
5
USD
76805-070
77602-198
77602-200
77627-014
Dimethyl succinate 99%
Dimethyl succinate
Precaución: for Research Use Only
Formula:
C₆H₁₀O₄ MW: 146.14 g/mol Boiling Pt: 200 |
MDL Number:
MFCD00008466 CAS Number: 106-65-0 Merck Index: 12,09038 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...