Indium(III) acetylacetonate 98%

Supplier: Ambeed
Danger

Synonyms: Indium (III) tris(2,4-pentanedionato-O,O'), Indium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')indium (III)

A512710-10G A512710-1G A512710-5G A512710-25G
76805-064EA 47 USD
76805-064 76805-066 76805-068 77602-196
Indium(III) acetylacetonate 98%
Indium(III) acetylacetonate

Precaución: for Research Use Only

Formula: C₁₅H₂₁InO₆
MW: 412.15 g/mol
MDL Number: MFCD00013494
CAS Number: 14405-45-9
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR