Diisopropylamine hydrochloride 97%

Supplier: Ambeed
Warning

A509485-25G A509485-5G
76805-054EA 30.77 USD
76805-054 76805-056
Diisopropylamine hydrochloride 97%
Diisopropylamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₅N
MW: 137.65 g/mol
MDL Number: MFCD00034860
CAS Number: 819-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR