Phe-Phe-OMe HCl 97%

Supplier: Ambeed
Warning

Synonyms: Methyl L-phenylalanyl-L-phenylalaninate hydrochloride

A508815-1G A508815-250MG A508815-5G
76805-042EA 54.11 USD
76805-042 76805-044 76805-046
Phe-Phe-OMe HCl 97%
Phe-Phe-OMe HCl

Precaución: for Research Use Only

Formula: C₁₉H₂₂N₂O₃·HCl
MW: 362.86 g/mol
MDL Number: MFCD00190986
CAS Number: 38017-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR