Methyl 2-(4-isobutylphenyl)propanoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-[4-(2-methylprop-1-yl)phenyl]propionate, Methyl2-(4-isobutylphenyl)propanoate, Methyl 2-[4-(2-methylprop-1-yl)phenyl]propanoate

A508535-1G A508535-25G A508535-5G
76805-036EA 16.98 USD
76805-036 76805-038 76805-040
Methyl 2-(4-isobutylphenyl)propanoate 97%
Methyl 2-(4-isobutylphenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₂
MW: 220.307 g/mol
MDL Number: MFCD00869922
CAS Number: 61566-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR