4-Bromo-8-methoxyquinoline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromoquinolin-8-yl methyl ether, 4-Bromo-8-methoxy-1-azanaphthalene
A507502-100MG
A507502-1G
A507502-250MG
76805-024EA
31.83
USD
76805-024
76805-026
76805-028
4-Bromo-8-methoxyquinoline 98%
4-Bromo-8-methoxyquinoline
Precaución: for Research Use Only
Formula:
C₁₀H₈BrNO MW: 238.081 g/mol |
MDL Number:
MFCD08063209 CAS Number: 103028-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...