MES sodium salt monohydrate 98%

Supplier: Ambeed
Warning

A507308-10G A507308-25G A507308-5G
76805-018EA 7 USD
76805-018 76805-020 76805-022
MES sodium salt monohydrate 98%
Formula: C₆H₁₂NNaO₄S.H₂O
MDL Number: MFCD31920063
CAS Number: 438589-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR