Methyl-1-methylcyclopropyl ketone 98%
Supplier: Ambeed
|
Danger
|
A506670-1G
A506670-5G
A506670-25G
A506670-10G
76805-014EA
20
USD
76805-014
76805-016
77602-170
77669-266
Methyl-1-methylcyclopropyl ketone 98%
Methyl-1-methylcyclopropyl ketone
Precaución: for Research Use Only
Formula:
C₆H₁₀O |
MDL Number:
MFCD00010245 CAS Number: 1567-75-5 UN: 1224 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...