Enalaprilat dihydrate 97%

Supplier: Ambeed
Warning

Synonyms: N-[(1S)-1-Carboxy-3-phenylpropyl]-L-alanyl-L-proline, Enalaprilic acid, (S)-1-((S)-2-(((S)-1-Carboxy-3-phenylpropyl)amino)propanoyl)pyrrolidine-2-carboxylic acid dihydrate

A502548-100MG A502548-10MG A502548-1MG A502548-250MG A502548-50MG
76804-994EA 79.57 USD
76804-994 76804-996 76804-998 76805-000 76805-002
Enalaprilat dihydrate 97%
Enalaprilat dihydrate

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₂O₅·2H₂O
MW: 348.40 g/mol
MDL Number: MFCD00941393
CAS Number: 84680-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR