Dimethylcyclopropane-1,1-dicarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 1,1-cyclopropanedicarboxylate, 1,1-Cyclopropanedicarboxylic acid dimethyl ester

A500494-100G A500494-1G A500494-25G A500494-500G A500494-5G
76804-976EA 91.24 USD
76804-976 76804-978 76804-980 76804-982 76804-984
Dimethylcyclopropane-1,1-dicarboxylate 95%
Dimethylcyclopropane-1,1-dicarboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀O₄
MW: 158.15 g/mol
MDL Number: MFCD00192079
CAS Number: 6914-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR