(1-Hexyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

Synonyms: Hexyltriphenylphosphonium bromide

A499731-25G A499731-500G A499731-100G
76804-968EA 8 USD
76804-968 76804-970 77551-518
(1-Hexyl)triphenylphosphonium bromide 97%
(1-Hexyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: H₃C(CH₂)₅P(C₆H₅)₃Br
MW: 427.36 g/mol
MDL Number: MFCD00011860
CAS Number: 4762-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR