Fmoc-D-Abu-OH 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-(Fmoc-amino)butyric acid

A498821-100G A498821-10G A498821-1G A498821-25G A498821-5G
76804-950EA 71 USD
76804-950 76804-952 76804-954 76804-956 76804-958
Fmoc-D-Abu-OH 97%
Fmoc-D-Abu-OH

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₄
MW: 325.364 g/mol
MDL Number: MFCD00270336
CAS Number: 170642-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR