Dioctyl ether 99% contains 100 ppm BHT as inhibitor

Supplier: Ambeed
Warning

Synonyms: n-Octyl ether, di-n-Octyl ether, Dioctylether

A497642-100G A497642-25G A497642-5G
76804-938EA 35 USD
76804-938 76804-940 76804-942
Dioctyl ether 99% contains 100 ppm BHT as inhibitor
Dioctyl ether

Precaución: for Research Use Only

Formula: C₁₆H₃₄O
MDL Number: MFCD00009563
CAS Number: 629-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR