Glutaryl dichloride 97%

Supplier: Ambeed
Danger

Synonyms: Glutaryl chloride, Glutaryl dichloride, Glutaroyl dichloride

A496866-25G A496866-5G
76804-934EA 43.5 USD
76804-934 76804-936
Glutaryl dichloride 97%
Glutaryl dichloride

Precaución: for Research Use Only

Formula: C₅H₆Cl₂O₂
MW: 169.0059 g/mol
MDL Number: MFCD00000755
CAS Number: 2873-74-7
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR