5-Phenyl-4-pentyn-1-ol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Phenylpent-4-yn-1-ol

A493111-1G A493111-250MG
76804-912EA 180.36 USD
76804-912 76804-914
5-Phenyl-4-pentyn-1-ol 98%
5-Phenyl-4-pentyn-1-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₂O
MW: 160.22 g/mol
MDL Number: MFCD00191481
CAS Number: 24595-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR