Ethyl-2-anilino-4-oxo-4,5-dihydro-3-thiophenecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-oxo-2-(phenylamino)-4,5-dihydrothiophene-3-carboxylate

A491535-1G A491535-5G A491535-100MG
76804-888EA 27 USD
76804-888 77594-536 77880-069
Ethyl-2-anilino-4-oxo-4,5-dihydro-3-thiophenecarboxylate 95%
Ethyl-2-anilino-4-oxo-4,5-dihydro-3-thiophenecarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₃S
MDL Number: MFCD02106374
CAS Number: 78267-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR