Fmoc-(R)-3-Cyclobutylalanine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclobutylpropanoic acid, Fmoc-D-β-Cyclobutyl-Ala-OH

A490886-100MG A490886-250MG A490886-5G
76804-882EA 9 USD
76804-882 76804-884 76804-886
Fmoc-(R)-3-Cyclobutylalanine 98%
Fmoc-(R)-3-Cyclobutylalanine

Precaución: for Research Use Only

Formula: C₂₂H₂₃NO₄
MDL Number: MFCD03840401
CAS Number: 478183-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR