Indanthrone 99%

Supplier: Ambeed
Warning

Synonyms: Dinaphtho[2,3-a:2',3'-h]phenazine-5,9,14,18(6H,15H)-tetraone

A490548-25G A490548-5G A490548-100G
76804-872EA 21.22 USD
76804-872 77602-120 77669-198
Indanthrone 99%
Indanthrone

Precaución: for Research Use Only

Formula: C₂₈H₁₄N₂O₄
MW: 442.43 g/mol
MDL Number: MFCD00046964
CAS Number: 81-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR