Dodecyl methacrylate 97% stabilised

Supplier: Ambeed
Danger

Synonyms: Dodecyl 2-methylacrylate, Lauryl methacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A489963-100G A489963-500G
76804-864EA 5.31 USD
76804-864 77880-065 77880-066
Dodecyl methacrylate 97% stabilised
Dodecyl methacrylate

Precaución: for Research Use Only

Formula: C₁₆H₃₀O₂
MW: 254.41 g/mol
MDL Number: MFCD00008972
CAS Number: 142-90-5
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR