n-Hexatriacontane 98+%

Supplier: Ambeed
Warning

Synonyms: Hexatriacontane

A481298-1G A481298-5G
76804-828EA 23.34 USD
76804-828 76804-830
n-Hexatriacontane 98+%
n-Hexatriacontane

Precaución: for Research Use Only

Formula: H₃C(CH₂)₃₄CH₃
MW: 506.98 g/mol
MDL Number: MFCD00009413
CAS Number: 630-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR