Diethyl (3-bromopropyl)phosphonate 96%

Supplier: Ambeed
Warning

Synonyms: (3-Bromopropyl)phosphonic Acid Diethyl Ester

A479825-1G A479825-25G A479825-5G A479825-100G
76804-822EA 9 USD
76804-822 76804-824 76804-826 77880-042
Diethyl (3-bromopropyl)phosphonate 96%
Diethyl (3-bromopropyl)phosphonate

Precaución: for Research Use Only

Formula: C₇H₁₆BrO₃P
MDL Number: MFCD00800966
CAS Number: 1186-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR