Dimethyl-2-oxo-4-phenylbutylphosphonate 95%

Supplier: Ambeed
Warning

Synonyms: Dimethyl (2-oxo-4-phenylbutyl)phosphonate

A477489-10G A477489-1G A477489-25G A477489-5G
76804-808EA 68.96 USD
76804-808 76804-810 76804-812 76804-814
Dimethyl-2-oxo-4-phenylbutylphosphonate 95%
Dimethyl-2-oxo-4-phenylbutylphosphonate

Precaución: for Research Use Only

Formula: C₁₂H₁₇O₄P
MW: 256.24 g/mol
MDL Number: MFCD02093040
CAS Number: 41162-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR