Ethyl docosanoate 95+%

Supplier: Ambeed
Warning

A476358-1G A476358-100MG A476358-250MG
76804-796EA 136 USD
76804-796 77880-035 77880-036
Ethyl docosanoate 95+%
Ethyl docosanoate

Precaución: for Research Use Only

Formula: C₂₄H₄₈O₂
MDL Number: MFCD00056275
CAS Number: 5908-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR