Hexamethylbenzene 98%

Supplier: Ambeed
Warning

A475869-1G A475869-25G A475869-5G
76804-790EA 14 USD
76804-790 76804-792 76804-794
Hexamethylbenzene 98%
Hexamethylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₈
MDL Number: MFCD00008523
CAS Number: 87-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR