Methyl 2-amino-4-methylnicotinate 95+%

Supplier: Ambeed
Warning

A474983-100MG A474983-250MG
76804-786EA 64 USD
76804-786 76804-788
Methyl 2-amino-4-methylnicotinate 95+%
Methyl 2-amino-4-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD09837641
CAS Number: 76336-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR