Dodecyl-3,4,5-trihydroxybenzoate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Gallic acid lauryl ester, n-Dodecyl gallate, Dodecyl 3,4,5-trihydroxybenzoate, Gallic acid dodecyl ester, Dodecyl gallate, Lauryl 3,4,5-trihydroxybenzoate, Lauryl gallate, Progalline LA
A474134-100G
A474134-10G
A474134-25G
A474134-500G
76804-780EA
46
USD
76804-780
76804-782
76804-784
77626-948
Dodecyl-3,4,5-trihydroxybenzoate 95%
Dodecyl-3,4,5-trihydroxybenzoate
Precaución: for Research Use Only
Formula:
C₁₉H₃₀O₅ MW: 338.44 g/mol |
MDL Number:
MFCD00002195 CAS Number: 1166-52-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...