3-Methyl-1-p-tolyl-5-pyrazolone 97%
Supplier: Ambeed
|
Warning
|
A474101-100G
A474101-25G
A474101-500G
A474101-5G
76804-772EA
21
USD
76804-772
76804-774
76804-776
76804-778
3-Methyl-1-p-tolyl-5-pyrazolone 97%
3-Methyl-1-p-tolyl-5-pyrazolone
Precaución: for Research Use Only
Formula:
C₁₁H₁₂N₂O |
MDL Number:
MFCD00035708 CAS Number: 86-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...