2-Hydroxyhexanoic acid 98%

Supplier: Ambeed
Warning

A472209-1G A472209-250MG A472209-5G
76804-756EA 40 USD
76804-756 76804-758 76804-760
2-Hydroxyhexanoic acid 98%
2-Hydroxyhexanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MW: 132.15 g/mol
MDL Number: MFCD00004584
CAS Number: 6064-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR