Inosine Pranobex 98%

Supplier: Ambeed
Warning

A470576-1G A470576-5G A470576-100G A470576-25G A470576-500G
76804-742EA 5 USD
76804-742 76804-744 77669-112 77669-114 77880-024
Inosine Pranobex 98%
Inosine Pranobex

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₄O₅·₃C₉H₉NO₃·3C₅H₁₃NO
MDL Number: MFCD05662374
CAS Number: 36703-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR