2-Hydroxy-3-(3-methylbut-2-enyl)-1,4-naphthoquinone 99%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-3-(3-methylbut-2-en-1-yl)naphthalene-1,4-dione, Lapachol

A467467-1MG A467467-250MG
76804-724EA 6.37 USD
76804-724 76804-726
2-Hydroxy-3-(3-methylbut-2-enyl)-1,4-naphthoquinone 99%
2-Hydroxy-3-(3-methylbut-2-enyl)-1,4-naphthoquinone

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.28 g/mol
MDL Number: MFCD00001679
CAS Number: 84-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR